Learn More
Essendant PAPERPRO MEDIA
Supplier:  Essendant EPSSP91203
Encompass
Epson Somerset has always been the first paper choice among serious artists and printmakers Ever since the 1700s and the days of handmade paper Somerset has enjoyed a reputation for quality that has continued to grow through the years Now Epson brings you this old-world paper with an inkjet coating to meet the quality requirements of the most demanding printmaker Somerset Velvet for Epson is a 100pct cotton acid-free paper that delivers long-lasting prints when printed with Epson Archival Ink or UltraChrome Ink Sheet/Roll Size- 24x50 ft Paper Grade (gsm)- 255 gsm Paper/Media Color(s)- White Color Family- White
Catalog No. 50-000-35414