Learn More
Essendant PAPERPROOFING24X100WE
Supplier:  Essendant EPSS042004
Encompass
Designed with 10 mil naturally bright white base Epson Proofing Paper White Semi-Matte is free of optical brighteners and ideally suited for proofing a wide variety of color critical commercial offset printing applications most commonly used in high-end sheet-fed printed products such as brochures collateral book covers and posters Coated with exacting standards Proofing Paper White Semi-Matte is specifically optimized for proofing applications using Epson UltraChrome K3 ink and displays outstanding short term stability Its weight base tint and wide color gamut makes this the perfect choice for commercial proofing applications Sheet/Roll Size- 24x100 ft Paper/Media Thickness (mil)- 10 mil Paper/Media Color(s)- White Color Family- White
Catalog No. 50-000-32312