Learn More
Essendant PAPERPROOFING36X100
Supplier:  Essendant EPSS045113
Encompass
Offering the widest color gamut available for accurate color reproductions this 9 mil version of our Standard Proofing Paper provides a base color heavyweight and gloss level designed to match color-critical commercial offset press applications Optimized for proofing applications when used with Epson UltraChrome K3 Ink this media delivers outstanding short-term stability In fact it s designed to meet the exacting standards established by the GRACoL Committee Instant Dry Sheet/Roll Size- 36x100 ft Paper/Media Thickness (mil)- 9 mil Paper/Media Color(s)- White Color Family- White
Catalog No. 50-000-35573