Learn More
ATCC Papilloma virus; genome; Purified plasmid DNA
Supplier:  ATCC 45151D
Encompass
This clone contains the full length viral genome of HPV-11 inserted into the BamHI site of pBR322. It was constructed by linearizing the virus at the unique BamHI site (nt 7072). Note: The depositor has requested that this deposit be distributed exclusively for scientific and not for commercial purposes.
Catalog No. 50-238-4415