Learn More
Genetex Inc PAPP A Monoclonal Antibody [MSVA-780M] HistoMAX™ (GTX04437) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04437
Encompass_Preferred
The PAPP A antibody [MSVA-780M] HistoMAX™ (GTX04437) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human placenta tissue using GTX04437 PAPP A antibody [MSVA-780M] HistoMAX. Strong papp A immunostaining of trophoblast cells of a first trimenon placenta.
Catalog No. NC3666951