Learn More
Selleck Chemical LLC paquinimod S9963-1G
SDP
Supplier:  Selleck Chemical LLC S99631G
Encompass_Preferred
Paquinimod (ABR 25757) a specific inhibitor of S100A8/A9 could rescue the pneumonia with substantial reduction of viral loads in SARS-CoV-2-infected mice
Catalog No. 50-000-23949