Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Paquinimod-d5-1 | 248282-01-1 | 98.2% | 355.44 g·mol-1 | C21H17D5N2O3 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY100442S15MG

Encompass_Preferred

Paquinimod-d5-1 is a deuterated analog of paquinimod intended for research use as an isotopically labeled reference standard. It serves as an S100A8/S100A9 inhibitor surrogate for analytical, tracer, and pharmacokinetic studies.

  • Isotopically labeled (d5) for tracer and mass spectrometry applications.
  • High listed purity: 98.2%.
  • Molecular weight 355.44 g·mol-1 for accurate dosing and calculations.
  • Supplied as a 5 mg solid suitable for small-scale experiments.
  • Molecular formula C21H17D5N2O3 with structural identifiers for verification.

Catalog No. 50-002-14060


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.