Learn More
Medchemexpress LLC Paquinimod-d5-1 | 98.2% | 355.44 | C21H17D5N2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100442S110MG
Encompass_Preferred
Paquinimod-d5-1 is a deuterium-labeled analogue of paquinimod used as a tracer and internal standard for quantitative analytical methods. It supports accurate measurement of paquinimod-related analytes in research workflows and is intended for laboratory use in method development and sample analysis.
  • Deuterium-labeled analog suitable for tracer and internal standard use.
  • Compatible with NMR, GC-MS, and LC-MS quantitative workflows.
  • High purity, 98.2%.
  • Molecular weight 355.44 and formula C21H17D5N2O3.
  • Supplied in small milligram quantities for analytical applications.
Catalog No. 50-002-14022