Learn More
Cayman Chemical PAR21-6mouserat 10mg
SDP
Supplier:  Cayman Chemical 2712510 MG
Encompass_Preferred
A peptide agonist of PAR2; induces relaxation in precontracted rat arteries; inhibits keratinocyte growth in the presence and absence of growth factors; inhibits LPS-induced pulmonary neutrophil influx and increases in MMP-2 activity in mice
Catalog No. 50-294-1197