Learn More
Medchemexpress LLC Para-iodoHoechst 33258 | 158013-43-5 | 98.9% | 534.39 g·mol^-1 | C25H23IN6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1563210MG
Encompass_Preferred
para-iodoHoechst 33258 is an iodinated analog of the Hoechst 33258 DNA-staining dye that binds the minor groove of double-stranded DNA and produces blue fluorescence used for nuclear labeling in microscopy.
  • Iodinated Hoechst analog that binds the minor groove of DNA and fluoresces blue.
  • Suitable for live-cell and fixed-cell nuclear staining in fluorescence microscopy.
  • High purity suitable for research applications (reported 98.9%).
  • Provided as a small research pack to enable method development and microscopy assays.
  • CAS number 158013-43-5 and molecular weight ~534.39 g·mol^-1 for unambiguous identification.
Catalog No. 50-002-88067