Learn More
Cayman Chemical -para-OctopamInhydrochl 5g
SDP
Supplier:  Cayman Chemical 142795 G
Encompass_Preferred
An endogenous biogenic amine commonly found in invertebrates; similar to norepinephrine in structure and action in invertebrates; considered a trace amine which may interfere with aminergic pathways in humans
Catalog No. 50-295-5347