Learn More
Electron Microscopy Sciences Paraffin Block Trimmer Wax Trimmer, 220 VAC 1/EA
SDP
Supplier:  Electron Microscopy Sciences 6285020
Encompass_Preferred
Our Wax Trimmer is an appliance for removing excess paraffin from embedding tissue cassettes. It is compact, tabletop design fits easily in any histology lab. Melted paraffin is shaved away. Collection tray stays in place by magnetic contact with the unit. The convenient PE disposable tray liner allows easy disposal of the runoff wax. No need to scrape out the collection tray. Multiple blocks may be trimmed simultaneously. Grooved surface for proper wax drainageTwo temperature settings – Low setting is approx. 80°F and high setting is 90°FUL listed power cordIncludes 5 pieces disposable drip tray linerDimensions: 11¼" (including tray) W x 5" D x 8" H (15 x 23 x 18 cm). 8 lbs (3.6kg)Electrical
Catalog No. 50-209-0728
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.