Learn More
Biochain Institute Inc Colon Tumor Matched Pair Tissue Array - 29 colon tumor/adjacent normal pairs. Plus positive control and negative control, 2 slides/PK
SDP
Supplier:  Biochain Institute Inc T8235790D2
Encompass_Preferred
BioChain's tissue arrays or tissue microarrays (TMAs) are a useful tool in biomarker research and drug target validation. It consists of paraffin blocks that are assembled in array fashion to allow multiplex histological analysis. BioChain's TMAs provide a valuable tool for analysis of protein, RNA, or DNA molecules in oncology, both the malignant and metastatic markers. TMAs can also be constructed with tissues from other diseases and species.
Catalog No. 50-181-866