Learn More
Biochain Institute Inc Multiple Tumor Tissue Array - 15 different tumors and their corresponding normal tissues spotted in duplicate, 2 slides/PK
SDP
Supplier:  Biochain Institute Inc T82357122
Encompass_Preferred
BioChain's tissue arrays or tissue microarrays (TMAs) are a useful tool in biomarker research and drug target validation. It consists of paraffin blocks that are assembled in array fashion to allow multiplex histological analysis. BioChain's TMAs provide a valuable tool for analysis of protein, RNA, or DNA molecules in oncology, both the malignant and metastatic markers. TMAs can also be constructed with tissues from other diseases and species.
Catalog No. 50-181-907