Learn More
Electron Microscopy Sciences Paraformaldehyde 1% with Glutaraldehyde 2% in 0.1M Phosphate Buffer, pH 7.4 10 ML
SDP
Supplier:  Electron Microscopy Sciences 1594930
Encompass_Preferred
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. Paraformaldehyde 1 % Glutaraldehyde 2% in 0.1M Phosphate Buffer. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
pH: 7.4
Catalog No. 50-190-1151