Learn More
AdipoGen Paramagnetoquinone A B
SDP
Supplier:  AdipoGen AGCN20315C500
Encompass_Preferred
Chemical. CAS 2090377-60-7 A, 2090377-61-8 B. Formula C22H12O9 A, C22H13NO8 B. MW 420.3 A419.3 B. Isolated from Actinoallomurus sp. Potent antibacterial agent. Shows MIC values lower than 0.015µg/ml against Gram-positive pathogens, including some antibiotic-resistant strains and selected Gram-negative bacteria.
Catalog No. 50-256-2609