Learn More
Sklar Instruments Z-Type Parametrium Scissors, Sklar 12 in., Blunt/Blunt, Slightly Curved, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA.
SDP
Supplier:  Sklar Instruments 164008
Encompass_Preferred
Z-Type Parametrium Scissors, Sklar. 12 in., Blunt/Blunt, Slightly Curved, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar Edge Tungsten Carbide Z-Type Parametrium Scissors are a non-ratcheted finger ring OB/GYN instrument used for cutting Parametrium tissue, also known as cervical stroma, in vaginal hysterectomy procedures. Z-Type Parametrium Scissors are available in multiple lengths with various curvatures so as to compliment the size and curvature of Z-Type Hysterectomy Forceps. This is so the two instruments can be used in conjunction with one another without blocking the view of the physician. Sklar Edge TSklar permanently bonds tungsten carbide inserts to the blades of the scissors which maximizes control and precision. Sklar Edge scissors can be identified by two gold coated finger rings. This product is slightly curved with smooth edges, blunt/blunt tips, and a length of 12 inches.
Catalog No. 50-118-1402