Learn More
Chem Service Inc Paraoxon,311-45-5,100MG
SDP
Supplier:  Chem Service Inc N12816100MG
Encompass_Preferred
Molecular Formula C10H14NO6P, Purity 98.10%, Molecular Weight 275.2, Boiling Point 420 °C, Synonyms: Diethyl 4-nitrophenylester phosphoric acid, Diethyl-p-nitrophenylphosphate, Parathion O-analog, Paraoxon-ethyl, Oxyparathion,
Catalog No. NC0811417