Learn More
Medchemexpress LLC Pararosaniline hydrochloride | 569-61-9 | MFCD00001657 | 97.3% | 323.83 g·mol⁻¹ | C19H18ClN3 | 25 G
SDP
Supplier:  Medchemexpress LLC HYW12777025G
Encompass_Preferred
Pararosaniline hydrochloride is a pH-responsive basic dye used as a biological stain and research reagent for staining proteins, cells, and tissues. It exhibits a maximum absorption at approximately 549 nm and is employed in histology, cytology, and various industrial dyeing applications.
  • Presents an absorption maximum at 549 nm.
  • pH-responsive basic dye suitable for staining proteins and cellular structures.
  • Reported purity of 97.3% for consistent experimental results.
  • Applicable to histology, cytology, and industrial dyeing processes.
  • Supplied in laboratory-scale pack sizes for research use.
Catalog No. 50-002-82627