Learn More
Medchemexpress LLC Parathyroid Hormone (1-34), bovine | 12583-68-5 | 99.9% | 4108.77 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP12525MG
Encompass_Preferred
Parathyroid Hormone (1-34), bovine is a potent agonist of the parathyroid hormone (PTH) receptor. It increases calcium and inorganic phosphate levels in vivo and can be used in research related to osteoporosis. It is for research use only and not sold to patients.
  • Potent parathyroid hormone (PTH) receptor agonist.
  • Increases calcium and inorganic phosphate levels in vivo.
  • Can be used for osteoporosis research.
  • Inhibits osteoblast proliferation in a dose-dependent manner (in vitro).
  • Induces diverse effects on calcium and phosphorus content of culture medium (in vitro).
  • Increases serum osteocalcin concentrations without changing serum inorganic phosphate or calcium concentrations in old animals (in vivo).
  • Significantly higher serum 1,25-dihydroxyvitamin D concentrations in PTH-treated senile female rats than controls (in vivo).
Catalog No. 50-004-23275