Learn More
Apexbio Technology LLC Parathyroid Hormone (1-34), bovine,1mg CAS# 12583-68-5
SDP
Supplier:  Apexbio Technology LLC A11141
Encompass_Preferred
Parathyroid hormone (PTH) is the most important endocrine regulator of calcium and phosphorus concentration in extracellular fluid, which is secreted by the chief cell of the parathyroid glands as a polypeptide containing 84 amino acids. It acts to increase the concentration of calcium (Ca2+) in the blood, whereas calcitonin (a hormone produced by the parafollicular cells (C cells) of the thyroid gland) acts to decrease calcium concentration.PTH acts to increase the concentration of calcium in the blood by acting upon the parathyroid hormone 1 receptor (high levels in bone and kidney) and the parathyroid hormone 2 receptor (high levels in the central nervous system, pancreas, testis, and placenta).PTH half-life is approximately 4 minutes. [1] Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0484