Learn More
CPC Scientific H-Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-OH 5MG
SDP
Catalog No. 502110336
Encompass
SEQUENCE: H-Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-OHONE-LETTER SEQUENCE: SVSEIQLMHNLGKHLNSMERVEWLRKKLQDVHNFMOLECULAR FORMULA: C181H291N55O51S2MOLECULAR WEIGHT: 4117.77STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [52232-67-4]SYNONYMS: pTH (1-34) (human), TeriparatideRESEARCH AREA: OsteoporosisREFERENCES:T. Takai et al., Peptide Chemistry, 1978, 187 (1979)G.W. Tregear et al., Hoppe-Seyler's Z. Physiol. Chem., 355, 415 (1974)
SKU(s): PTHP-002A, PTHP-002B, PTHP-002C
Catalog No. 50-211-0336 Supplier CPC Scientific Supplier No. PTHP002C
Add to Cart
Add to Cart
SEQUENCE: H-Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-OHONE-LETTER SEQUENCE: SVSEIQLMHNLGKHLNSMERVEWLRKKLQDVHNFMOLECULAR FORMULA: C181H291N55O51S2MOLECULAR WEIGHT: 4117.77STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [52232-67-4]SYNONYMS: pTH (1-34) (human), TeriparatideRESEARCH AREA: OsteoporosisREFERENCES:T. Takai et al., Peptide Chemistry, 1978, 187 (1979)G.W. Tregear et al., Hoppe-Seyler's Z. Physiol. Chem., 355, 415 (1974)
SKU(s): PTHP-002A, PTHP-002B, PTHP-002C