Learn More
Medchemexpress LLC Paraxanthine-13C4,15N3 | 1173018-79-5 | MFCD11656179 | 99.8% | 187.12 g/mol | C7H8N4O2 | 1 MG

Supplier: Medchemexpress LLC HYW016498S11MG
Isotopically labeled paraxanthine (13C4,15N3) intended for use as a tracer and internal standard for quantitative analytical methods. Supplied as a white to light yellow solid with high purity for research use only. Powder should be stored at -20°C; solutions are stable at -80°C for up to 6 months or -20°C for up to 1 month.
- Isotopically labeled for accurate tracer studies and quantitation.
- High purity (99.8%) suitable for analytical standards.
- Solid powder form for easy handling and aliquoting.
- Soluble in DMSO and DMF; limited solubility in ethanol.
- Recommended storage conditions provided for long-term stability.
- Compatible with NMR, GC-MS, and LC-MS workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.