Learn More
Medchemexpress LLC HY-W016498 25mg Medchemexpress, Paraxanthine CAS:611-59-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01649825MG
Encompass_Preferred
Medchemexpress, HY-W016498 25mg Paraxanthine CAS:611-59-6 Paraxanthine, a caffeine metabolite, provides protection against Dopaminergic cell death via stimulation of Ryanodine Receptor Channels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3395