Learn More
Medchemexpress LLC Paraxanthine-d6 | 117490-41-2 | 98.9% | 186.20 g/mol | C7H2D6N4O2 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYW016498S500UG
Encompass_Preferred
Paraxanthine-d6 is the deuterium-labeled analogue of paraxanthine supplied as a stable-isotope internal standard for analytical and research applications. It is intended for research use only and is commonly used in LC-MS and GC-MS quantification of caffeine metabolites and related metabolic and pharmacokinetic studies.
  • Deuterium-labeled internal standard for LC-MS and GC-MS.
  • High isotopic enrichment to support accurate quantification.
  • Suitable for pharmacokinetic and metabolic studies of caffeine and its metabolites.
  • Available in small analytical pack sizes for trace-level assays.
  • For research use only; not for human or clinical use.
Catalog No. 50-003-03323