Learn More
Selleck Chemical LLC Pardoprunox S0104-5MG
SDP
Supplier:  Selleck Chemical LLC S01045MG
Encompass_Preferred
Pardoprunox hydrochloride (DU-126891 SME-308) is a potent but partial dopamine D2 receptor agonist with pEC50 of 8 0 and a partial agonist in the induction of [35S]GTP S binding with pEC50 of 9 2 and a serotonin 5-HT1A receptor agonist with pEC50 of 6 3 respectively
Catalog No. 50-000-41013