Learn More
Apexbio Technology LLC Parecoxib Sodium 198470-85-8 25mg
SDP
Supplier:  Apexbio Technology LLC B123925
Encompass_Preferred
Parecoxib sodium (CAS 198470-85-8) is a small-molecule inhibitor that selectively targets cyclooxygenase-2 (COX-2) an enzyme involved in the biosynthesis of pro-inflammatory prostaglandins By preferentially blocking COX-2 activity while sparing COX-1 parecoxib sodium modulates inflammatory pathways with reduced effects on physiological prostaglandin function This compound is widely utilized in research settings to investigate COX-2 mediated inflammatory processes pain mechanisms and the pharmacological modulation of prostaglandin-driven pathways
Catalog No. 50-000-90396