Learn More
Selleck Chemical LLC Parecoxib Sodium S5418-25mg
SDP
Supplier:  Selleck Chemical LLC S541825MG
Encompass_Preferred
Parecoxib Sodium (SC-69124) is a water-soluble injectable sodium salt form of parecoxib which is a potent and selective inhibitor of cyclo-oxygenase-2 (COX2)
Catalog No. 50-310-9962