Learn More
ATCC Aspergillus flavus; var. flavus
Supplier:  ATCC 22546
Encompass
Major corn pathogen that causes Aspergillus ear rot. This strain was isolated in 1971 from moldy corn in Iowa, United States.
Catalog No. 50-238-0772
 Restricted Item