Learn More
Medchemexpress LLC Pargyline hydrochloride | 306-07-0 | 99.9% | C11H14ClN | 1 ML
SDP
Supplier:  Medchemexpress LLC HYA009110MM/1ML
Encompass_Preferred
Pargyline hydrochloride is an irreversible monoamine oxidase (MAO) inhibitor with Kis of 13 μM for MAO-A and 0.5 μM for MAO-B. It exhibits antihypertensive and anticancer activities. Pargyline (hydrochloride) is also a click chemistry reagent, containing an Alkyne group, which can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have Azide groups.
  • Irreversible monoamine oxidase (MAO) inhibitor
  • Inhibits MAO-A with a Ki of 13 μM
  • Inhibits MAO-B with a Ki of 0.5 μM
  • Demonstrates antihypertensive activities
  • Exhibits anticancer activities
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
Catalog No. 50-004-33560