Learn More
Medchemexpress LLC Pargyline hydrochloride | 306-07-0 | MFCD00012492 | 99.91% | C11H14ClN | 100 MG
SDP
Supplier:  Medchemexpress LLC HYA0091100MG
Encompass_Preferred
Pargyline hydrochloride is an irreversible monoamine oxidase (MAO) inhibitor with antihypertensive and anticancer activities. It functions as a click chemistry reagent, containing an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Irreversible monoamine oxidase (MAO) inhibitor
  • Exhibits antihypertensive activities
  • Demonstrates anticancer activities
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
Catalog No. 50-004-32115