Learn More
Medchemexpress LLC Paricalcitol-d6 | 2070009-67-3 | 99.9% | 422.67 g/mol | C27H38D6O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY7658510MG
Encompass_Preferred
Paricalcitol-d6 is the deuterium-labeled analogue of paricalcitol provided as a high-purity research standard for use as a tracer and internal standard in quantitative analytical workflows.
  • Deuterium-labeled internal standard for LC-MS and GC-MS.
  • Suitable as an NMR reference and tracer in metabolic studies.
  • High purity for accurate quantitation (99.85%).
  • Supplied as a small-scale powder; store cold to preserve stability.
Catalog No. 50-002-98156