Learn More
Medchemexpress LLC HY-109098 5mg Medchemexpress, Parimifasor CAS:1796641-10-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090985MG
Encompass_Preferred
Medchemexpress, HY-109098 5mg Parimifasor CAS:1796641-10-5 Parimifasor (LYC30937) is an immunomodulator, with anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8678