Learn More
Selleck Chemical LLC Paritaprevir (ABT-450) S5404-5mg
SDP
Supplier:  Selleck Chemical LLC S54045MG
Encompass_Preferred
Paritaprevir (ABT-450) is a nonstructural (NS) protein 3/4A protease inhibitor
Catalog No. 50-310-9911