Learn More
Selleck Chemical LLC Paromomycin Sulfate S4183-1g
SDP
Supplier:  Selleck Chemical LLC S41831G
Encompass_Preferred
Paromomycin Sulfate(Aminosidine sulfate) is an aminoglycoside antibiotics inhibiting protein synthesis in non-resistant cells by binding to 16S ribosomal RNA
Catalog No. 50-310-8644