Learn More
American Radiolabeled Chemicals Inc PAROXETINE 3H 25UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 177725UC
Encompass
Paroxetine-phenyl-6'-3H 25uCi in ethanol, 1 mCi/ml, S.A. 15-30 Ci/mmol, M.W. 329.4, Shipped in Dry Ice
Catalog No. 50-901-117