Learn More
Medchemexpress LLC Paroxetine | 61869-08-7 | 99.9% | 1 ML

Supplier: Medchemexpress LLC HY12227210MM 1ML
Paroxetine is a selective serotonin reuptake inhibitor (SSRI) supplied as a ready-to-use 10 mM solution in DMSO for research applications. The product is provided as a 1 mL vial and intended for laboratory research use only.
- Ready-to-use 10 mM solution in DMSO.
- Supplied as a 1 mL aliquot for small-scale experiments.
- High reported purity (99.9%).
- CAS number 61869-08-7 for compound identification.
- Molecular formula C19H20FNO3 and molecular weight 329.37 g·mol⁻1.
- For research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.