Learn More
Abcam Paro x etine hydrochloride, 5-HT uptake inhibitor, 10MG
Supplier:  Abcam AB12006910MG
Encompass_Preferred
MW 365.8 Da, Purity >99%. Potent, selective 5-HT (serotonin) uptake inhibitor. Inhibits human and rat 5-HT transporters (Ki values are 0.065 and 0.05 nM, respectively). Antidepressant and anxiolytic in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7887