Learn More
STEMCELL Technologies ME0328, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 1000271
Encompass
ME0328 is a cell-permeable and selective inhibitor of poly(ADP-ribose) polymerase 3 (PARP-3; IC₅₀ = 0.89 µM; Lindgren et al.). PARP-3 is required for stabilization of the mitotic spindle and crucial for mitotic progression (Sharif-Askari et al.). CANCER RESEARCH· Enhances vinorelbine-induced microtubule destabilization, leading to cell cycle arrest and apoptosis in breast cancer cells (Sharif-Askari et al.).
Catalog No. 50-204-5515
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov