Learn More
Medchemexpress LLC PARP1-IN-11 | 2482484-87-5 | 99.4% | 296.28 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY147886100MG
Encompass_Preferred
PARP1-IN-11 (compound 49) is a potent PARP1 inhibitor with an IC50 value of 0.082 μM. It demonstrates complete inhibition of PARP2 and substantially inhibits PARP3, TNKS1, and TNKS2.
  • Potent PARP1 inhibitor
  • IC50 value of 0.082 μM for PARP1
  • Completely inhibits PARP2
  • Substantially inhibits PARP3
  • Substantially inhibits TNKS1
  • Substantially inhibits TNKS2
Catalog No. 50-003-63088