Learn More
TARGETMOL CHEMICALS INC PARP1-IN-5 dihydrochloride 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T96105MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. PARP1-IN-5 dihydrochloride is an orally active, potent and selective PARP-1 inhibitor (IC50 =14.7 nM). PARP1-IN-5 dihydrochloride can be used for the research of cancer. Purity 96.03%
Catalog No. 50-288-0194