Learn More
Medchemexpress LLC Parp1-in-8 | 836640-15-4 | 99.3% | 403.86 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147030
Encompass_Preferred
PARP1-IN-8 is a potent and BBB-penetrated PARP1 inhibitor with an IC50 of 97 nM. It demonstrates significant anti-proliferative activity against human lung adenocarcinoma epithelial cell line A549, and does not display significant cytotoxicity on HFF cells.
  • Potent, BBB-penetrated PARP1 inhibitor
  • IC50 of 97 nM
  • Significantly potent anti-proliferative activity against A549 cells
  • No significant cytotoxicity on HFF cells
Catalog No. 50-003-63291