Learn More
Medchemexpress LLC Parp10/15-In-2 | 2892064-99-0 | >=95% | 286.26 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY146501
Encompass_Preferred
PARP10/15-IN-2 (Compound 8h) is a potent PARP10 and PARP15 dual inhibitor. It can enter cells and rescue them from apoptosis.
  • Potent PARP10 and PARP15 dual inhibitor
  • IC50 values of 0.15 μM against PARP10
  • IC50 values of 0.37 μM against PARP15
  • Able to enter cells
  • Rescues cells from apoptosis
Catalog No. 50-003-63484