Learn More
Medchemexpress LLC HY-109068A 5mg Medchemexpress, Parsaclisib (hydrochloride) CAS:1995889-48-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY109068A5MG
Encompass_Preferred
Medchemexpress, HY-109068A 5mg Parsaclisib (hydrochloride) CAS:1995889-48-9 Parsaclisib hydrochloride (INCB050465 hydrochloride) is a potent, selective and orally active inhibitor of PI3Kδ, with an IC50 of 1 nM at 1 mM ATP. Parsaclisib hydrochloride shows approximately 20000-fold selectivity over other PI3K class I isoforms. Parsaclisib hydrochloride can be used for the research of relapsed or refractory B-cell malignancies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0762