Learn More
Grainger Parts Washer Solvent 1 Gal.
Supplier:  Grainger 16W543
Encompass_Preferred
The Blaster Parts Washer Cleaner Solvent is a heavy-duty cleaner and degreaser designed for use in parts washers. It quickly dissolves grease, oil, tar, wax, dirt, moisture, and other contaminants from parts and surfaces. This non-flammable formula is safe on metal, plastic, rubber and painted surfaces. It contains no SARA 313 reportable ingredients. This one gallon container of Blaster solvent is made in the USA.
Catalog No. 50-276-3772