Learn More
A-M Systems PARYLENE-C INSULATED MICROELEC
SDP
Supplier:  A-M Systems 573400
Encompass_Preferred
Parylene-C Insulated Microelectrodes feature a Tungsten substrate with a 12-degree tip.
Catalog No. NC2226265