Learn More
Diagnostic Biosystems PAS PAS
Supplier:  Diagnostic Biosystems KT027
Encompass_Preferred
The Periodic Acid Schiff (PAS) Stain Kit is intended for use in histological demonstration of lymphocytes and mucopolysaccharides. The staining pattern of the lymphocytes are helpful in making therapeutic decisions in established cases of lymphocytic leukemia. The PAS reaction in tissue sections is useful for the demonstration of mucopolysaccharides. PAS staining may also be used for the demonstration of fungal organisms in tissue sections.
Catalog No. 50-240-7501