Learn More
Medchemexpress LLC Pasireotide acetate | 396091-76-2 | 99.3% | 1107.26 | C60H70N10O11 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16381A10MG
Encompass_Preferred
Pasireotide acetate is a somatostatin receptor agonist peptide supplied as the acetate salt for research and preclinical studies. It is commonly used to study hormone secretion and endocrine disease models, and is provided with supporting analytical data such as COA, HNMR, and LCMS.
  • Somatostatin receptor agonist peptide for research use
  • Acetate salt form, suitable for aqueous preparations
  • High purity (~99.3%) as reported by manufacturer
  • Available in multiple pack sizes, including 10 MG
  • Used in studies of GH, IGF-I, and ACTH secretion and endocrine disease models
Catalog No. 50-002-88592