Learn More
Medchemexpress LLC Pasireotide acetate | 396091-76-2 | 99.5% | 1107.26 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY16381A5MG
Encompass_Preferred
Pasireotide acetate, also known as SOM230 acetate, is a long-acting cyclohexapeptide somatostatin analogue. It exhibits improved agonist activity across multiple somatostatin receptor subtypes (sst1, sst2, sst3, and sst5) and has demonstrated the ability to suppress the secretion of growth hormone (GH), insulin-like growth factor-I (IGF-I), and adrenocorticotropic hormone (ACTH), suggesting its potential effectiveness in managing conditions like acromegaly and Cushing's disease. Additionally, it possesses antisecretory, antiproliferative, and proapoptotic activities.
  • Improved agonist activity at somatostatin receptors (sst1/2/3/5)
  • Suppresses GH, IGF-I, and ACTH secretion
  • Potential efficacy in acromegaly and Cushing's disease
  • Exhibits antisecretory activity
  • Exhibits antiproliferative activity
  • Exhibits proapoptotic activity
Catalog No. 50-003-93673