Learn More
Medchemexpress LLC Pasireotide acetate | 396091-76-2 | 99.5% | 1107.26 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16381A50MG
Encompass_Preferred
Pasireotide acetate is a long-acting cyclohexapeptide somatostatin analogue that improves agonist activity at somatostatin receptors. It can suppress GH, IGF-I, and ACTH secretion, indicating potential efficacy in acromegaly and Cushing's disease. This compound also exhibits antisecretory, antiproliferative, and proapoptotic activity.
  • Improves agonist activity at somatostatin receptors (subtypes sst1/2/3/4/5, pKi=8.2/9.0/9.1/<7.0/9.9, respectively).
  • Suppresses GH, IGF-I, and ACTH secretion, indicating potential efficacy in acromegaly and Cushing's disease.
  • Exhibits antisecretory, antiproliferative, and proapoptotic activity.
Catalog No. 50-003-93697